missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium bromide, 99.8% (metals basis)
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
Description
Rubidium bromide is used in infrared optics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1340°C |
| Melting Point | 693°C |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Merck Index | 14,8283 |
| Solubility Information | Soluble in water |
| Formula Weight | 165.38 |
| Percent Purity | 99.8% |
| Odor | Odorless |
| Refractive Index | 1.553 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction