missing translation for 'onlineSavingsMsg'
Learn More

Rubidium carbonate, 99.8% (metals basis)

Product Code. 11330719
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11330719

Brand: Thermo Scientific Alfa Aesar 010562.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 584-09-8 | CO3Rb2 | 230.94 g/mol

Rubidium carbonate is used in glass-making by enhancing stability and durability as well as reducing its conductivity. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas. Rubidium carbonate glass has been extensively tested for use in anti-collision devices for motor vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-09-8
Molecular Formula CO3Rb2
Molecular Weight (g/mol) 230.94
MDL Number MFCD00011188
InChI Key WPFGFHJALYCVMO-UHFFFAOYSA-L
Synonym rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate
PubChem CID 11431
IUPAC Name rubidium(1+);carbonate
SMILES [Rb+].[Rb+].[O-]C([O-])=O

Specifications

Melting Point 837°C
Color White
Physical Form Powder
Quantity 50 g
Linear Formula Rb2CO3
Solubility Information Soluble in water.
Formula Weight 230.95
Percent Purity 99.8%
Odor Odorless
Sensitivity Very Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.