missing translation for 'onlineSavingsMsg'
Learn More

Rubidium carbonate, 99%, pure

Product Code. p-2692139 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10115620

Brand: Thermo Scientific Acros 317150250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 584-09-8 | CO3Rb2 | 230.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-09-8
Molecular Formula CO3Rb2
Molecular Weight (g/mol) 230.94
MDL Number MFCD00011188
InChI Key WPFGFHJALYCVMO-UHFFFAOYSA-L
Synonym rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate
PubChem CID 11431
IUPAC Name rubidium(1+);carbonate
SMILES [Rb+].[Rb+].[O-]C([O-])=O

Specifications

Melting Point 837°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 98.5% min. (ex Rb)
Linear Formula Rb2CO3
Solubility Information Solubility in water: 4500g/L (20°C)
Formula Weight 230.95
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Rubidium carbonate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.