missing translation for 'onlineSavingsMsg'
Learn More
Rubidium carbonate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals Product Code.: 11346128

Rubidium carbonate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

Product Code. 11346128
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11346128

Brand: Thermo Scientific Alfa Aesar 011105.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It finds it application as a raw material for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 584-09-8
Molecular Formula CO3Rb2
Molecular Weight (g/mol) 230.94
MDL Number MFCD00011188
InChI Key WPFGFHJALYCVMO-UHFFFAOYSA-L
Synonym rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonateShow MoreShow Less
PubChem CID 11431
SMILES [Rb+].[Rb+].[O-]C([O-])=O

Especificaciones

Melting Point 837°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.975% (metals basis)
Solubility Information Soluble in water at 20°C 4500g/L.
IUPAC Name dirubidium(1+) carbonate
Formula Weight 230.95
Odor Odorless
Sensitivity Very Hygroscopic
Appearance White powder
Chemical Name or Material Rubidium carbonate
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY28D031 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number011105.03,011105.14,011105.06
Lot Number10253190 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number011105.03,011105.14,011105.06
Lot NumberU14K006 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number011105.03,011105.14,011105.06
Lot NumberM24I014 Certificate TypeCertificate of Analysis Date22/11/2022 Catalog Number011105.03,011105.14,011105.06
Lot NumberF14X025 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011105.03,011105.14,011105.06

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Rubidium carbonate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado