missing translation for 'onlineSavingsMsg'
Learn More
Rubidium chloride, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11350719

Rubidium chloride, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11350719
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11350719

Brand: Thermo Scientific Alfa Aesar 010563.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubidium chloride is the mostly used rubidium compound, and finds use in various fields ranging from electrochemistry to molecular biology. It is an efficient non-invasive biomarker; increases dopamine and norepinephrine levels, and useful for anergic and apathetic depressives. Pharmaceutical compositions have been used as antidepressants in Europe. It is employed in biochemistry to induce cells to take up DNA. This compound increases the competence of the cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-11-9
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+);chloride
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390°C
Melting Point 718°C
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.8% (metals basis)
Linear Formula RbCl
Merck Index 14,8284
Solubility Information Soluble in water. Sparingly soluble in methanol.
Formula Weight 120.92
Percent Purity 99.8%
Odor Odorless
Refractive Index 1.493
Sensitivity Hygroscopic
Density 2.800 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX10K040 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number010563.18,010563.09
Lot NumberA0462188 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number010563.18,010563.09
Lot NumberU14K024 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number010563.18,010563.09
Lot NumberR28K002 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number010563.18,010563.09
Lot NumberP21K001 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number010563.18,010563.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium chloride, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.