missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, 99.8+%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 11984191
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-11-9
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390.0°C
Melting Point 718.0°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99.8+%
Linear Formula RbCl
Merck Index 15,8417
Solubility Information Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water, practically insoluble in alcohols
IUPAC Name rubidium(1+) chloride
Formula Weight 120.92
Percent Purity ≥99.8%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Rubidium chloride
Show More Show Less
Product Identifier
  • Rubidium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'