missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. p-4229018 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11934191

Brand: Thermo Scientific Acros 193920100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390.0°C
Melting Point 718.0°C
Color White
Physical Form Chunks and/or Powder
Quantity 10 g
Assay Percent Range 28.9 to 30.1% (Cl) (Argentometry)
Linear Formula RbCl
Merck Index 15,8417
Solubility Information Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water, practically insoluble in alcohols
IUPAC Name rubidium(1+) chloride
Formula Weight 120.92
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Rubidium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.