missing translation for 'onlineSavingsMsg'
Learn More
Rubidium chloride, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 15421027

Rubidium chloride, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 15421027
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15421027

Brand: Thermo Scientific Alfa Aesar 012892.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Testing for perchloric acid, source of rubidium, catalyst

It is used in testing for perchloric acid in laboratories, and is a source of rubidium. Rubidium chloride is used as a catalyst and additive in petrol. It has been employed to prepare molecular nanowires for nanoscale devices. Thin layer of rubidium chloride enhance the attributes of organic light-emitting diodes. Rubidium chloride also plays an essential role for enhancement in the memory storage property. It is an excellent non-invasive biomarker, and used in biomedical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-11-9
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chlorideShow MoreShow Less
PubChem CID 62683
ChEBI CHEBI:78672
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390°C
Melting Point 718°C
Color White
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99% (metals basis)
Merck Index 14,8284
Solubility Information Soluble in water; Practically insoluble in alcohol.
IUPAC Name rubidium(1+);chloride
Formula Weight 120.92
Percent Purity 99%
Odor Odorless
Refractive Index 1.493
Sensitivity Hygroscopic
Density 2.800 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU14K024 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number012892.14,012892.06,012892.22
Lot NumberR28K002 Certificate TypeCertificate of Analysis Date29/07/2024 Catalog Number012892.14,012892.06,012892.22
Lot NumberP21K001 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number012892.14,012892.06,012892.22
Lot NumberP07I040 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number012892.14,012892.06,012892.22
Lot NumberR23I052 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number012892.14,012892.06,012892.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium chloride, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.