missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium chloride, 99+%, Thermo Scientific Chemicals
£154.77 - £392.97
Chemical Identifiers
CAS | 7791-11-9 |
---|---|
Molecular Formula | ClRb |
Molecular Weight (g/mol) | 120.92 |
MDL Number | MFCD00011187 |
InChI Key | FGDZQCVHDSGLHJ-UHFFFAOYSA-M |
Synonym | rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chlorideShow More |
PubChem CID | 62683 |
ChEBI | CHEBI:78672 |
SMILES | [Cl-].[Rb+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10042800
|
Thermo Scientific Acros
201270250 |
25 g | Glass bottle |
£154.77
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657164
|
Thermo Scientific Acros
201271000 |
100 g | Glass bottle |
£392.97
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7791-11-9 | |
120.92 | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
62683 | |
[Cl-].[Rb+] |
ClRb | |
MFCD00011187 | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chlorideShow More | |
CHEBI:78672 |
Specifications
1390.0°C | |
7791-11-9 | |
Crystalline Powder and/or Crystals | |
ClRb | |
MFCD00011187 | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
rubidium(1+);chloride | |
62683 | |
120.92 | |
2.8g/mL |
718.0°C | |
White | |
99+% | |
RbCl | |
15, 8417 | |
Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water, practically insoluble in alcohols | |
[Cl-].[Rb+] | |
120.92 | |
CHEBI:78672 | |
≥99% | |
Rubidium chloride |
Safety and Handling
missing translation for 'einecsNumber' : 232-240-9
Product Identifier
- Rubidium chloride
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rubidium chloride, 99+%, Thermo Scientific Chemicals