missing translation for 'onlineSavingsMsg'
Learn More

Rubidium fluoride, 99.1% (metals basis), Thermo Scientific Chemicals

£46.53 - £187.94

Chemical Identifiers

CAS 13446-74-7
Molecular Formula FRb
Molecular Weight (g/mol) 104.466
MDL Number MFCD00011191
InChI Key AHLATJUETSFVIM-UHFFFAOYSA-M
Synonym rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
PubChem CID 83473
IUPAC Name rubidium(1+);fluoride
SMILES [F-].[Rb+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492867
View Documents
Thermo Scientific Alfa Aesar
047144.09
10 g
£46.53
10g
Please to purchase this item. Need a web account? Register with us today!
15402877
View Documents
Thermo Scientific Alfa Aesar
047144.18
50 g
£187.94
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of catalyst and for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13446-74-7
104.466
AHLATJUETSFVIM-UHFFFAOYSA-M
83473
[F-].[Rb+]
FRb
MFCD00011191
rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
rubidium(1+);fluoride

Specifications

795°C
White
FRb
MFCD00011191
rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
AHLATJUETSFVIM-UHFFFAOYSA-M
rubidium(1+);fluoride
83473
99.1%
Hygroscopic
(metals basis)
13446-74-7
Crystalline
RbF
UN3288
Soluble in water. (1306g/L at 20°C).
[F-].[Rb+]
104.466
104.47
1.398
3.557 g/mL
Rubidium fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 236-603-2

missing translation for 'rtecsNumber' : VL8740000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Rubidium fluoride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Rubidium fluoride (RbF)

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only