missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium fluoride hydrate, 99%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Raw material used in the perparation of rubidium metal.
Solubility
Soluble in water.
Notes
Store in a cool, dry, conditions in well sealed container.
Raw material used in the perparation of rubidium metal.
Solubility
Soluble in water.
Notes
Store in a cool, dry, conditions in well sealed container.

Specifications
Specifications
Quantity | 50 g |
UN Number | UN3288 |
Solubility Information | Soluble in water. |
Formula Weight | 104.47 (Anhydrous) |
Percent Purity | 99% |
Odor | Odorless |
Chemical Name or Material | Rubidium fluoride hydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rubidium fluoride hydrate, 99%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction