missing translation for 'onlineSavingsMsg'
Learn More
Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis), Thermo Scientific Chemicals Product Code.: 11335799

Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis), Thermo Scientific Chemicals

Product Code. 11335799
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11335799

Brand: Thermo Scientific Alfa Aesar 010565.NC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubidium Hydroxide is used to give fireworks a violet color. It is a notable ntermediate in the synthesis of other rubidium compounds. It is used in photography and storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-82-3
Molecular Formula HORb
Molecular Weight (g/mol) 102.48
MDL Number MFCD00011194
InChI Key CPRMKOQKXYSDML-UHFFFAOYSA-M
Synonym rubidium hydroxide, rubidium hydroxide rb oh, rboh, rubidium 1+ hydroxide, rubidium hydroxide solution, rubidiumol, rubidium hydoxide, rubidium hydroxide sup 81 rb, rubidium monohydroxide, rb.hoShow MoreShow Less
PubChem CID 62393
ChEBI CHEBI:32108
IUPAC Name rubidium(1+);hydroxide
SMILES [OH-].[Rb+]

Specifications

Color Yellow
Physical Form Liquid
Quantity 10 g
Linear Formula RbOH
UN Number UN2677
Merck Index 14,8285
Solubility Information Soluble in ethanol and water.
Formula Weight 102.48 (Anhydrous)
Percent Purity ≥99.6%
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Assay (metals basis)
Chemical Name or Material Rubidium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP08K049 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog Number010565.NC,010565.NE
Lot NumberX30J028 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number010565.NC,010565.NE
Lot NumberW11J039 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number010565.NC,010565.NE
Lot NumberQ12J004 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number010565.NC,010565.NE
Lot NumberY03I035 Certificate TypeCertificate of Analysis Date09/11/2022 Catalog Number010565.NC,010565.NE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.