missing translation for 'onlineSavingsMsg'
Learn More
Rubidium iodide, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11356868

Rubidium iodide, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11356868
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11356868

Brand: Thermo Scientific Alfa Aesar 013497.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubidium iodide is used for making metal rubidium and other rubidium salts. It is also used in the production of special glass, miniature high energy battery and crystal scintillation counter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-29-6
Molecular Formula IRb
Molecular Weight (g/mol) 212.372
MDL Number MFCD00011192
InChI Key WFUBYPSJBBQSOU-UHFFFAOYSA-M
Synonym rubidium iodide, rubidium-iodid, rubidium jodatum, unii-18cyw0vl2x, rubidium 1+ iodide, rubidium 1+ ion iodide, 18cyw0vl2x, i.rb, acmc-20ajs7Show MoreShow Less
PubChem CID 3423208
IUPAC Name rubidium(1+);iodide
SMILES [Rb+].[I-]

Specifications

Boiling Point 1300°C
Melting Point 647°C
Color White
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in water,alcohol and alkali aqueous solution.
Formula Weight 212.37
Odor Odorless
Refractive Index 1.6474
Sensitivity Hygroscopic
Appearance White crystalline
Density 3.55
Chemical Name or Material Rubidium iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR21EE059 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number013497.18,013497.09
Lot NumberU06K029 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number013497.18,013497.09
Lot NumberR10K028 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number013497.18,013497.09
Lot NumberZ12J062 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number013497.18,013497.09
Lot NumberR12J025 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number013497.18,013497.09,013497

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium iodide, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.