missing translation for 'onlineSavingsMsg'
Learn More

Rubidium iodide, 99.8% (metals basis)

Product Code. p-4407779
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11366868

Brand: Thermo Scientific Alfa Aesar 013497.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7790-29-6 | IRb | 212.372 g/mol

Rubidium iodide is used for making metal rubidium and other rubidium salts. It is also used in the production of special glass, miniature high energy battery and crystal scintillation counter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-29-6
Molecular Formula IRb
Molecular Weight (g/mol) 212.372
MDL Number MFCD00011192
InChI Key WFUBYPSJBBQSOU-UHFFFAOYSA-M
Synonym rubidium iodide, rubidium-iodid, rubidium jodatum, unii-18cyw0vl2x, rubidium 1+ iodide, rubidium 1+ ion iodide, 18cyw0vl2x, i.rb, acmc-20ajs7
PubChem CID 3423208
IUPAC Name rubidium(1+);iodide
SMILES [Rb+].[I-]

Specifications

Boiling Point 1300°C
Melting Point 647°C
Color White
Physical Form Crystals
Quantity 10 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in water,alcohol and alkali aqueous solution.
Formula Weight 212.37
Odor Odorless
Refractive Index 1.6474
Sensitivity Hygroscopic
Appearance White crystalline
Density 3.55 g/cm3
Chemical Name or Material Rubidium iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.