Learn More
Rubidium nitrate, 99%, Thermo Scientific Chemicals
£51.56 - £751.74
Chemical Identifiers
CAS | 13126-12-0 |
---|---|
Molecular Formula | NO3Rb |
Molecular Weight (g/mol) | 147.47 |
MDL Number | MFCD00011193 |
InChI Key | RTHYXYOJKHGZJT-UHFFFAOYSA-N |
Synonym | rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrateShow More |
PubChem CID | 25731 |
IUPAC Name | rubidium(1+);nitrate |
SMILES | [Rb+].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387617
|
Thermo Scientific Alfa Aesar
012889.09 |
10 g |
£51.56
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317627
|
Thermo Scientific Alfa Aesar
012889.18 |
50 g |
£197.55
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397617
|
Thermo Scientific Alfa Aesar
012889.30 |
250 g |
£751.74
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rubidium nitrate used in infrared radiation producing pyrotechnic compositions as a colorant and an oxidizer. It is also used as a raw material for preparation of other rubidium compounds and rubidium metal, for manufacture of catalysts and in scintillation counters. It is rarely used in fireworks to produce a red-violet colour.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13126-12-0 | |
147.47 | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
25731 | |
[Rb+].[O-][N+]([O-])=O |
NO3Rb | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrateShow More | |
rubidium(1+);nitrate |
Specifications
310°C | |
White | |
NO3Rb | |
UN1477 | |
Soluble in water. Slightly soluble in acetone. Very soluble in nitric acid. | |
[Rb+].[O-][N+]([O-])=O | |
147.47 | |
147.47 | |
1.52 | |
3.11 g/mL | |
Rubidium nitrate |
13126-12-0 | |
Crystalline | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
rubidium(1+);nitrate | |
25731 | |
99.8% | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P210-P220-P221-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335-H350
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 236-060-1
missing translation for 'rtecsNumber' : QV0900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.