missing translation for 'onlineSavingsMsg'
Learn More
Rubidium perchlorate, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11341728

Rubidium perchlorate, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11341728
50 g
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11341728

Brand: Thermo Scientific Alfa Aesar 011628.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A perchlorate oxidizing agent. Rubidium perchlorate as chosen because its precipitation provides an excellent separation from strontium and since it is also an accepted weighing form for rubidium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-42-4
Molecular Formula ClO4Rb
Molecular Weight (g/mol) 184.914
MDL Number MFCD00016294
InChI Key NQYGGOOZLKJKPS-UHFFFAOYSA-M
Synonym rubidium perchlorate, rubidium 1+ ion perchlorate, perchloric acid, rubidium salt, rubidiumperchlorat, perchloric acid, rubidium salt 1:1, clo4.rb, rubidium chlorate vii, acmc-20ajs4, perchloric acid rubidium salt, rubidium 1+ perchlorate ion
PubChem CID 23673707
IUPAC Name rubidium(1+);perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Rb+]

Specifications

Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Solubility Information Soluble in water (180mg/ml at 100°C).
Formula Weight 184.94
Percent Purity 99.5%
Odor Odorless
Refractive Index 1.4701
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium perchlorate, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium perchlorate, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.