missing translation for 'onlineSavingsMsg'
Learn More

Rubidium sulfate, 99% (metals basis)

Product Code. 11326868
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11326868

Brand: Thermo Scientific Alfa Aesar 013493.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7488-54-2 | H2O4RbS | 183.54 g/mol

Rubidium sulfate is used in hematology stain. It is also used as the raw materials for making kinds of Rubidium salts. It is fused on a ceramic bead and used as thermoionic detector. It is also used as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7488-54-2
Molecular Formula H2O4RbS
Molecular Weight (g/mol) 183.54
MDL Number MFCD00011190
InChI Key XNNUAZGCOYTNQE-UHFFFAOYSA-N
PubChem CID 197088
IUPAC Name sulfuric acid rubidium
SMILES [Rb].OS(O)(=O)=O

Specifications

Melting Point 1050°C
Physical Form Crystalline
Quantity 100 g
Synonym rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate
Solubility Information Soluble in water (424mg/ml at 10°C).
Formula Weight 267
Percent Purity 99%
Odor Odorless
Refractive Index 1.513
Density 3.613 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium sulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.