Learn More
Fisherbrand™ runVIEW™ Mini and Midi Horizontal Gel System Packages PROMO
runVIEW is designed for real-time size fractionation and recovery of nucleic acids. runVIEW can maximise efficiency of DNA recovery from EtBr and SYBR stained gels by minimising number of steps involved in post-electrophoretic purification.
Brand:  Fisherbrand™ F-RVBSBV-LID-MIDI
 View more versions of this product
Product Code. 15301367
  • £341.00 / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Portable adjustable blue light illuminator viewing platform can be used with SUB-GEL Mini and SUB-GEL Midi electrophoresis tanks
  • bluVIEW emission filter lid with built-in extractor fan enables condensation free viewing of blue light fluorescently stained 70mm or 100mm wide agarose gels
  • Blue light is completely safe and results in improved cloning efficiency compared to UV
Three types of packages available:
  • runVIEW™ CONVERTER package includes runVIEW™ adjustable viewing platform and bluVIEW Mini or Midi emission filter lid (allows standard SUB-GEL Mini or SUB-GEL Midi tanks to be converted to real-time electrophoresis)
  • runVIEW™ STANDARD package includes runVIEW™ adjustable viewing platform and runVIEW™ SUB-GEL Mini or Midi tank
  • runVIEW™ COMPLETE package includes STANDARD package in Mini or Midi format with power supply
Specifications
Specifications
Applications: Education - ideal for quick checks of low and medium numbers of samples following PCR and cloning
CONVERTER Package
runVIEWTM viewing platform and bluVIEW lid (for use with Sub-Gel Midi Horizontal Gel Electrophoresis Unit)