Learn More
Novus Biologicals™ RUNX1 Plasmid
Mammalian expression vector construct, pCMV/hRUNX1, containing coding sequence of human Runt-related transcription factor 1
Brand: Novus Biologicals™ NBP2-25281-100UG
Additional Details : Weight : 0.00970kg
Description
The RUNX1 Plasmid is a mammalian expression vector construct, pCMV/hRUNX1, containing coding sequence of human Runt-related transcription factor 1 (GenBank Accession: BC136381). Gene ID: 861Specifications
0.5mg/mL | |
Expression of hRUNX1 protein in mammalian cells. Induction of FOXP3 promoter activity. Total content of 10μg is enough for one 96-well cell transfection (0.1μg plasmid per well). Recommended amount for other plate formats: 6-well plate (2μg/well), 12-well plate (0.8μg/well) and 24-well plate (0.4μg/well). | |
100 μg |
Store at -20C. Avoid freeze-thaw cycles. | |
RUNX1 Plasmid - Target | |
Human (Hu) |