missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium, 0.5% on 3 mm alumina tablets, Thermo Scientific™
CO or CO2 from H2 by methanation to CH4
£84.09 - £276.00
Chemical Identifiers
CAS | 7440-18-8 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
Synonym | on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon |
PubChem CID | 23950 |
ChEBI | CHEBI:30682 |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
23950 | |
ruthenium |
Ru | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbonShow More | |
CHEBI:30682 | |
[Ru] |
Specifications
7440-18-8 | |
Ru | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
Ruthenium, 0.5% on alumina tablets, 3mm alumina tablets |
Tablets | |
MFCD00011207 MFCD03458417 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : TSCA
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ruthenium, 0.5% on 3 mm alumina tablets, Thermo Scientific™