missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, 0.5% on granular carbon

Product Code. 11304457
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
missing translation for 'unitSize'
100g
25g
This item is not returnable. View return policy

Product Code. 11304457

missing translation for 'mfr': Thermo Scientific Alfa Aesar 038332.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-18-8 | Ru | 101.07 g/mol

Ruthenium is used in wear-resistant electrical contacts and the production of thick-film resistors. Its use in some platinum alloys, and as a catalyst. It is a most effective hardeners for platinum and palladium. It is also used in some advanced high-temperature single-crystal super alloys, with applications including the turbine blades in jet engines and fountain pen nibs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Quantity 100 g
UN Number UN1325
Solubility Information Soluble in fused alkalis. Insoluble in water,acid and aqua regia.
Concentration or Composition (by Analyte or Components) 0.5% on granular carbon
Odor Odorless
Chemical Name or Material Ruthenium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.