missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium, 4% on 6.35mm (0.25in) alumina rings, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
As Heterogeneous Catalysts. It is suitable for use as an electrocatalyst in gas diffusion electrodes. As Electrochemical gas sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Gray-White |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Ruthenium, 4% on 6.35mm (0.25 in.) alumina rings |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'