missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium on activated carbon, 5% Ru, unreduced, ca. 50% moisture, Thermo Scientific Chemicals Product Code.: 10521261

Ruthenium on activated carbon, 5% Ru, unreduced, ca. 50% moisture, Thermo Scientific Chemicals

Product Code. 10521261
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10521261

Brand: Thermo Scientific Acros 195460250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-18-8, 7440-44-0, 7732-18-5
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Color Black
Density 1.02
Quantity 25 g
Fieser 01,983; 05,574
Merck Index 15, 8433
Formula Weight 101.07
Percent Purity 5% (Ru on dry basis)(typically)
Packaging Glass bottle
Form Powder
Chemical Name or Material Ruthenium on activated carbon, 5% Ruthenium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0336072 Certificate TypeCertificate of Analysis Date09/12/2020 Catalog Number195461000,195460250
Lot NumberA0335264 Certificate TypeCertificate of Analysis Date20/11/2020 Catalog Number195461000,195460250
Product Identifier
  • Ruthenium on carbon, ca. 50% moisture

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium on activated carbon, 5% Ru, unreduced, ca. 50% moisture, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.