missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium(III) 2,4-pentanedionate, Ru 24% min, Thermo Scientific Chemicals Product Code.: 11300729

Ruthenium(III) 2,4-pentanedionate, Ru 24% min, Thermo Scientific Chemicals

Product Code. 11300729
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11300729

Brand: Thermo Scientific Alfa Aesar 010568.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Hydrogenation

Precursor for the preparation of other compounds of ruthenium. Ruthenium(III) 2,4-pentanedionate is employed as a recyclable catalyst for certain organic transformations like the acetylation of phenols, alcohols, and amines under neat conditions. It is also used as a homogeneous catalyst, for example, in the hydrolysis of sodium borohydride and regiospecific tritiation of aromatic carboxylic acids. It is also used as a catalyst for enantioselective hydrogenation of acids such as aryl acrylic acid and aryl propenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-93-6
Molecular Formula C15H21O6Ru
Molecular Weight (g/mol) 398.40
MDL Number MFCD00000030
InChI Key RTZYCRSRNSTRGC-LNTINUHCSA-K
Synonym ruthenium acetylacetonate, ruthenium iii acetylacetonate, acetylacetone ruthenium iii salt, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-8ci, z-4-oxopent-2-en-2-olate; ruthenium 3+Show MoreShow Less
PubChem CID 5488829
SMILES [Ru+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 260°C (decomposition)
Color White
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in most organic solvents such as acetone,chlorinated hydrocarbons,alcohols,cyclohexane and benzene. Insoluble in water.
IUPAC Name (Z)-4-oxopent-2-en-2-olate;ruthenium(3+)
Formula Weight 398.4
Appearance Red crystals
Chemical Name or Material Ruthenium(III) 2, 4-pentanedionate, Ru 24% min.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP22J002 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number010568.06,010568.03
Lot NumberN18G013 Certificate TypeCertificate of Analysis Date25/10/2022 Catalog Number010568.06,010568.03,010579.22
Lot Number10238201 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010568.06,010568.03,010579.22
Lot NumberT27D047 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010568.06,010568.03,010579.22
Lot NumberNZ0010 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010568.06,010568.03,010579.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium(III) 2,4-pentanedionate, Ru 24% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.