missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous

Product Code. p-4509792 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 10541071

Brand: Thermo Scientific Acros 197210020

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-08-8 | Cl3Ru | 207.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-08-8
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.43

Specifications

Melting Point >500.0°C
Color Black
Physical Form Crystals or Powder
Quantity 2 g
Assay Percent Range 48 to 50% (Ru)
Linear Formula RuCl3
Fieser 04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292
Merck Index 15,8436
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water, slightly soluble in alcohol
Formula Weight 207.43
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ruthenium(III) chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ruthenium(III) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.