Learn More
Ruthenium(III) chloride, anhydrous, Ru 47.7% min, Thermo Scientific Chemicals
Description
Ruthenium(III) chloride is widely used as a starting material of ruthenium complexes. It acts as a catalyst used in the oxidative cyclization of 1,7-dienes to oxepane diols. It is used in the hydroxylation of tertiary hydrocarbons in combination of periodate or bromate. It is involved as a catalyst in the synthesis of 2,3-pyridinedicarboxylic acid-13C3, 15N, where the unlabelled analog is an inhibitor of glucose synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >500°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 10 g |
Linear Formula | RuCl3 |
UN Number | UN3260 |
Merck Index | 14,8302 |
Formula Weight | 207.43 |
Concentration or Composition (by Analyte or Components) | Ru 47.7% min |
Odor | Pungent |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P25L003 | Certificate of Analysis | 27/03/2025 | 047182.04,047182.09 |
X30K027 | Certificate of Analysis | 07/11/2024 | 047182.04,047182.09 |
U07K060 | Certificate of Analysis | 21/08/2024 | 047182.04,047182.09 |
S13K050 | Certificate of Analysis | 09/07/2024 | 047182.04,047182.09 |
R09F021 | Certificate of Analysis | 08/12/2021 | 047184.03,047182.04,047182.09 |
Safety and Handling



- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.