missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium(III) chloride hydrate, 35 - 40% Ru, Thermo Scientific Chemicals Product Code.: 13418448

Ruthenium(III) chloride hydrate, 35 - 40% Ru, Thermo Scientific Chemicals

Product Code. 13418448
100 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 13418448

Brand: Thermo Scientific Acros 195481000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-67-0
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.42
MDL Number MFCD00149844
InChI Key YBCAZPLXEGKKFM-UHFFFAOYSA-K
Synonym Ruthenium trichloride
IUPAC Name ruthenium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Color Black to Brown
Physical Form Powder
Quantity 100 g
Linear Formula RuCl3·xH2O
Fieser 04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292
Merck Index 14, 8302
Solubility Information Solubility in water: soluble
Formula Weight 207.42
Percent Purity 35 to 40% Ru
Packaging Glass bottle
Chemical Name or Material Ruthenium(III) chloride hydrate, 35 - 40% Ru
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0446163 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number195480250,195480050,195480010,195481000
Lot NumberA0455128 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number195480250,195480050,195480010,195481000
Lot NumberA0399609 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number195480250,195480050,195480010,195481000
Lot NumberA0404128 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number195480250,195480050,195480010,195481000
Lot NumberA0405433 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number195480250,195480050,195480010,195481000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ruthenium(III) chloride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium(III) chloride hydrate, 35 - 40% Ru, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.