missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Thermo Scientific Chemicals Product Code.: 11391678

Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Thermo Scientific Chemicals

Product Code. 11391678
50 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11391678

Brand: Thermo Scientific Alfa Aesar 011043.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxidation.Ruthenium(III) chloride hydrate is used as a catalyst for alkyne functionalization and oxidation reactions. It is used as key raw materials for the preparation of various ruthenium-based catalysts, including Grubbs’ catalysts. It is utilized in the preparation of nanoparticulate ruthenium-aluminum oxyhydroxide catalyst, which is efficient for dehydrogenation of alcohols to the corresponding aldehydes. It is also involved in the synthesis of 2-ethyl-3-methylquinolines from primary aromatic amines and triallylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-67-0
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.42
MDL Number MFCD00149844
InChI Key YBCAZPLXEGKKFM-UHFFFAOYSA-K
IUPAC Name ruthenium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Melting Point 100°dec
Physical Form Crystalline Soluble
Quantity 50 g
Assay Percent Range 99.9%
Solubility Information Very soluble in water. Soluble in alcohol,acetone.
Formula Weight 207.43 (Anhydrous)
Odor Pungent
Sensitivity Hygroscopic
Appearance Black crystalline or powder
Chemical Name or Material Ruthenium(III) chloride hydrate, PGM basis, Ru 38% min.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX11K024 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number011043.09,011043.18,011043.04
Lot NumberM25K024 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number011043.09,011043.18,011043.04
Lot NumberM04K025 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog Number011043.09,011043.18,011043.04
Lot NumberT24J072 Certificate TypeCertificate of Analysis Date14/08/2023 Catalog Number011043.09,011043.18,011043.04
Lot NumberQ21J010 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog Number011043.09,011043.18,011043.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.