missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium(III) iodide, anhydrous, Ru 20.5% min, Thermo Scientific Chemicals Product Code.: 11311848

Ruthenium(III) iodide, anhydrous, Ru 20.5% min, Thermo Scientific Chemicals

Product Code. 11311848
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11311848

Brand: Thermo Scientific Alfa Aesar 011805.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13896-65-6
MDL Number MFCD00016316
Synonym ruthenium iii iodide, ruthenium iii ; iodide, ruthenium 3+ triiodide, ruthenium iii iodide anhydrous, ruthenium iii iodide, anhydrous 1g, ruthenium iii iodide, anhydrous, ru minShow MoreShow Less

Specifications

Melting Point 590°C (decomposition)
Physical Form Powder
Quantity 1 g
Formula Weight 481.78
Concentration or Composition (by Analyte or Components) Ru 20.5% min.
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) iodide, Anhydrous

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN07K032 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog Number011805.03,011805.06
Lot NumberT13J010 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number011805.03,011805.06
Lot NumberP22J011 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number011805.03
Lot NumberX05I017 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number011805.03,011806.06,011805.06
Lot NumberP10E025 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011805.03,011806.06,011805.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium(III) iodide, anhydrous, Ru 20.5% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.