missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution

Product Code. 10793403 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10793403

Brand: Thermo Scientific Acros 365040250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34513-98-9 | N4O10Ru | 317.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34513-98-9
Molecular Formula N4O10Ru
Molecular Weight (g/mol) 317.09
MDL Number MFCD00016313

Specifications

Color Brown to Orange
Physical Form Solution
Quantity 25 mL
Solubility Information Solubility in water: soluble
Formula Weight 317.09
Percent Purity 1.5% (Ru) (typically)
Packaging Glass bottle
Specific Gravity 1.07
Density 1.0700g/mL
Chemical Name or Material Ruthenium(III) nitrosyl nitrate, In diluted nitric acid
compliance-icons
Product Identifier
  • Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.