missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylnitrate, Ru 31.3% min

Product Code. p-4824912
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11366558

Brand: Thermo Scientific Alfa Aesar 012175.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34513-98-9 | N4O10Ru

Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34513-98-9
Molecular Formula N4O10Ru
MDL Number MFCD00016313

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Assay Percent Range Ru 31.3% min.
Solubility Information Miscible with cold water.
Formula Weight 317.09
Odor Odorless
Appearance Red-brown crystals
Chemical Name or Material Ruthenium(III) nitrosylnitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.