missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide, 99.5+%, anhydrous

Product Code. p-221826 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10523771

Brand: Thermo Scientific 218340010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12036-10-1 | O2Ru | 133.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-10-1
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00011210
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
Synonym ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
PubChem CID 82848
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O

Specifications

Color Black
Physical Form Fine Powder
Quantity 1 g
Assay Percent Range 99.5+%
Linear Formula RuO2
Fieser 12,428; 14,272; 15,281
Solubility Information Solubility in water: insoluble
Formula Weight 133.07
Percent Purity ≥99.5%
Packaging Glass bottle
Specific Gravity 6.97
Density 6.9700g/mL
Chemical Name or Material Ruthenium(IV) oxide, 99.5+%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.