Learn More
Ruthenium(IV) oxide, anhydrous, 99.9%, Thermo Scientific Chemicals
Description
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Applied as an electrochemical super capacitor material, has high capacity to store charge.
- Used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, used as active material in super capacitors.
- Dispersion property of RuO2; used to increase the capacitance of nanocomposite electrode materials for supercapacitors
- Extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries

Specifications
Specifications
Boiling Point | 1200°C (sublimation) |
CAS | 12036-10-1 |
Color | Black |
Physical Form | Powder |
Quantity | 25 g |
Molecular Formula | O2Ru |
MDL Number | MFCD00011210 |
UN Number | UN1479 |
Synonym | ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous |
Solubility Information | Not soluble in water |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10249767 | Certificate of Analysis | 22/02/2024 | A10816.03,A10816.14,A10816.06 |
10247701 | Certificate of Analysis | 15/08/2023 | A10816.03,A10816.14,A10816.06 |
10245212 | Certificate of Analysis | 05/01/2023 | A10816.03,A10816.14,A10816.06 |
10237864 | Certificate of Analysis | 12/04/2022 | A10816.03,A10816.14,A10816.06 |
X22A017 | Certificate of Analysis | 22/11/2021 | A10816.03,A10816.14,A10816.06 |
Safety and Handling

- Danger
- Oxidising solid Category 2
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.