missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide hydrate, > 54% Ru, Thermo Scientific Chemicals

£58.69 - £1385.90

Chemical Identifiers

CAS 32740-79-7
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00149846
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10276920
View Documents
Thermo Scientific Acros
197430010
1 g Glass bottle
£58.69
1g
Please to purchase this item. Need a web account? Register with us today!
10582791
View Documents
Thermo Scientific Acros
197430050
5 g Glass bottle
£293.46
5g
Please to purchase this item. Need a web account? Register with us today!
10741994
View Documents
Thermo Scientific Acros
197430250
25 g Glass bottle
£1385.90
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32740-79-7
133.07
WOCIAKWEIIZHES-UHFFFAOYSA-N
O=[Ru]=O
O2Ru
MFCD00149846
dioxoruthenium

Specifications

32740-79-7
Fine Powder
RuO2·xH2O
01,984; 14,272; 04,108; 05,161
WOCIAKWEIIZHES-UHFFFAOYSA-N
dioxoruthenium
133.07
Ruthenium(IV) oxide hydrate, >54% Ru
Black
O2Ru
MFCD00149846
Solubility in water: insoluble
O=[Ru]=O
133.07
>54% (Ru)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2

Hazard Statement
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only