missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals Product Code.: 11315687

Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals

Product Code. 11315687
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11315687

Brand: Thermo Scientific Alfa Aesar 043403.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical catalyst, important material for producing resistance and capacitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32740-79-7
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00149846
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O

Specifications

Melting Point 1200°C (sublimation)
Color Black
Physical Form Powder
Quantity 5 g
UN Number UN1479
Solubility Information It is soluble in fused alkalis. It is insoluble in water.
Formula Weight 133.07 (Anhydrous)
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Ru 54 to 60%
Assay (metals basis)
Chemical Name or Material Ruthenium (IV) oxide hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25K004 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog Number043403.03,043403.14,043403.06
Lot NumberY05E011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043403.14,043403.06,043392.18,043392.30
Lot NumberT16B026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043403.14,043403.06,043392.18,043392.30
Lot NumberW26D039 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number043403.14,043403.06,043392.18,043392.30
Lot NumberK15L34 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number043403.14,043403.06,043392.18,043392.30
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.