Learn More
Thermo Scientific Chemicals Ruthenium Red, ≥85%, pure
£95.10 - £322.00
Chemical Identifiers
CAS | 11103-72-3 |
---|---|
Molecular Formula | H24Cl6N14O2Ru3 |
Molecular Weight (g/mol) | 786.35 |
InChI Key | JQJSTVUROJELSR-UHFFFAOYSA-H |
Synonym | 6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride |
PubChem CID | 16218584 |
IUPAC Name | azane;ruthenium(2+);hexachloride;dihydrate |
SMILES | N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15519343
|
Thermo Scientific Chemicals
279710010 |
1 g |
£95.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15529343
|
Thermo Scientific Chemicals
279710050 |
5 g |
£322.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
11103-72-3 | |
786.35 | |
6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride | |
azane;ruthenium(2+);hexachloride;dihydrate |
H24Cl6N14O2Ru3 | |
JQJSTVUROJELSR-UHFFFAOYSA-H | |
16218584 | |
N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2] |
Specifications
> 500.0°C | |
≥85% | |
[(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6 | |
6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride | |
JQJSTVUROJELSR-UHFFFAOYSA-H | |
azane;ruthenium(2+);hexachloride;dihydrate | |
16218584 | |
≥85% | |
Glass Bottle |
11103-72-3 | |
H24Cl6N14O2Ru3 | |
15,8434 | |
Solubility in water: soluble. | |
N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2] | |
786.35 | |
786.35 | |
Pure | |
Ruthenium Red |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
- Ruthenium Red
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P362-Take off contaminated clothing.
RUO – Research Use Only