missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium sponge, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11316418

Ruthenium sponge, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11316418
50 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11316418

Brand: Thermo Scientific Alfa Aesar 012062.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

UN Number UN3089
Quantity 50 g
Percent Purity 99.95%
Mesh Size −20 Mesh
Odor Odorless
Form Sponge
Assay (metals basis)
Chemical Name or Material Ruthenium sponge

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP29I027 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012062.09,012062.18,012062.04,012062.SS
Lot NumberA09L10 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012062.09,012062.18,012062.04,012062.SS
Lot NumberJ28X012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012062.09,012062.18,012062.04,012062.SS
Lot NumberC12N30 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012062.09,012062.18,012062.04,012062.SS
Lot NumberZ07C074 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012062.09,012062.18,012062.04,012062.SS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium sponge, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.