missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£251.00 - £310.00
Chemical Identifiers
CAS | 7440-18-8, 7647-01-0, 7732-18-5 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
PubChem CID | 23950 |
ChEBI | CHEBI:30682 |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10108693
|
SPEX Certiprep
PLRU3-2Y |
125 mL | Triple-leached LDPE bottle |
£251.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10749234
|
SPEX Certiprep
PLRU3-2X |
500 mL | Triple-leached LDPE bottle |
£310.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ru | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-18-8, 7647-01-0, 7732-18-5 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
CHEBI:30682 | |
[Ru] |
Ru | |
MFCD00011207 MFCD03458417 | |
23950 | |
ruthenium |
Specifications
7440-18-8, 7647-01-0, 7732-18-5 | |
0.1, 10.0, 89.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Ru | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
Assurance | |
10% HCl |
0.1, 10.0, 89.9 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011207 MFCD03458417 | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
Matrix Concentration Cat. No. 10% HCl 1000μg/mL PLRU3-2Y, -2X 10,000 PLRU3-3Y, -3X |
|
Ruthenium Standard |
Safety and Handling
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1000
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local, regional, national and international regulations.
Product intended for laboratory use only