Learn More
Ruthenium standard solution, for AAS, 1mg/mL Ru in 5% HCl, Thermo Scientific Chemicals
£28.60 - £94.30
Specifications
Chemical Name or Material | Ruthenium standard solution |
---|---|
Concentration | 990 to 1010μg/mL of Ru(3+) in 10−20% HCl |
Name Note | 1 mg/ml Ru in 5% HCl |
CAS | 7440-18-8, 7647-01-0, 7732-18-5 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10266440
|
Thermo Scientific Acros
196251000 |
100 mL | Plastic bottle |
£28.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964081
|
Thermo Scientific Acros
196255000 |
500 mL | Plastic bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Ruthenium standard solution | |
1 mg/ml Ru in 5% HCl | |
Plastic bottle | |
Ru | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
101.07 | |
1.0200g/mL | |
107.0°C | |
Soluble in water |
990 to 1010μg/mL of Ru(3+) in 10−20% HCl | |
7440-18-8, 7647-01-0, 7732-18-5 | |
Yellow to Brown | |
MFCD00011207 MFCD03458417 | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
AAS | |
-25.0°C | |
1.02 | |
Solution |
Safety and Handling
GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection.
Danger
- Ruthenium atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only