missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(1-Naphthyl)ethanol, 97%, Thermo Scientific™

Product Code. 15416578
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15914-84-8
Melting Point 47°C to 49°C
Color Yellow
Molecular Formula C12H12O
Linear Formula C10H7CH(CH3)OH
MDL Number MFCD00077831
Quantity 1 g
Beilstein 2045009
Synonym s---1-1-naphthyl ethanol, s-1-naphthalen-1-yl-ethanol, s-1-naphthalen-1-yl ethanol, s---1-1-naphtyl ethanol, 1s-1-naphthalen-1-yl ethan-1-ol, s-1-1-naphthyl ethanol, s---alpha-methyl-1-naphthalenemethanol, 1s-1-naphthalen-1-ylethanol, 1s-1-naphthalen-1-yl ethanol, s-+-alpha-methyl-1-naphthalenemethanol
InChI Key CDRQOYRPWJULJN-VIFPVBQESA-N
SMILES CC(C1=CC=CC2=CC=CC=C21)O
IUPAC Name (1S)-1-naphthalen-1-ylethanol
Molecular Weight (g/mol) 172.227
PubChem CID 6976566
Formula Weight 172.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'