missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-(2-Chlorophenyl)ethanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
It is an intermediate in organic syntheses and used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate in organic syntheses and used as a pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room tempera
It is an intermediate in organic syntheses and used as a pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room tempera
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 131864-71-6 |
Quantity | 1 g |
Molecular Formula | C8H9ClO |
MDL Number | MFCD06659506 |
Synonym | s-1-2-chlorophenyl ethanol, 1s-1-2-chlorophenyl ethan-1-ol, 1s-1-2-chlorophenyl ethanol, s-1-2-chlorophenyl ethan-1-ol, pubchem5670, s-1-2-chloro-phenyl-ethanol, 5-1-2-chloro-phenyl-ethanol, s-2-chloro-alpha-methylbenzyl alcohol, benzenemethanol,2-chloro-a-methyl-, as |
Solubility Information | Slightly soluble in water. |
InChI Key | DDUBOVLGCYUYFX-LURJTMIESA-N |
SMILES | CC(C1=CC=CC=C1Cl)O |
IUPAC Name | (1S)-1-(2-chlorophenyl)ethanol |
Molecular Weight (g/mol) | 156.609 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'