missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-1-(4-Bromophenyl)ethylamine, ChiPros, 99%, ee 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(S)-(-)-1-(4-Bromophenyl)ethylamine is utilized as an important raw material in organic synthesis. It also serves as an intermediate in organic synthesis and pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-1-(4-Bromophenyl)ethylamine is utilized as an important raw material in organic synthesis. It also serves as an intermediate in organic synthesis and pharmaceuticals and agrochemicals.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place.
(S)-(-)-1-(4-Bromophenyl)ethylamine is utilized as an important raw material in organic synthesis. It also serves as an intermediate in organic synthesis and pharmaceuticals and agrochemicals.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -25°C (neat) |
Density | 1.371 |
Boiling Point | 140°C to 145°C (30 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.567 |
Quantity | 1 g |
UN Number | UN2735 |
Beilstein | 5729988 |
Sensitivity | Air Sensitive |
IUPAC Name | (1S)-1-(4-bromophenyl)ethanamine |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'