missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1-Benzyl-3-aminopyrrolidine, 99%, Thermo Scientific™
£79.00 - £282.00
Chemical Identifiers
CAS | 114715-38-7 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.26 |
InChI Key | HBVNLKQGRZPGRP-NSHDSACASA-N |
Synonym | s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine |
PubChem CID | 1519353 |
IUPAC Name | (3S)-1-benzylpyrrolidin-3-amine |
SMILES | C1CN(CC1N)CC2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10881691
|
Thermo Scientific Acros
437880010 |
1 g | Glass bottle |
£79.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10483520
|
Thermo Scientific Acros
437880050 |
5 g | Glass bottle |
£282.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
114715-38-7 | |
176.26 | |
s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine | |
(3S)-1-benzylpyrrolidin-3-amine |
C11H16N2 | |
HBVNLKQGRZPGRP-NSHDSACASA-N | |
1519353 | |
C1CN(CC1N)CC2=CC=CC=C2 |
Specifications
114715-38-7 | |
100.0 | |
1.0200g/mL | |
140°C | |
98.5% min. (GC) | |
1.02 | |
s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine | |
C1CN(CC1N)CC2=CC=CC=C2 | |
176.26 | |
176.26 | |
Liquid |
98.5 | |
Colorless to Yellow | |
108°C to 115°C (5.0 mmHg) | |
Authentic | |
C11H16N2 | |
+ 2 | |
HBVNLKQGRZPGRP-NSHDSACASA-N | |
(3S)-1-benzylpyrrolidin-3-amine | |
1519353 | |
99% | |
(S)-(+)-1-Benzyl-3-aminopyrrolidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (S)-(+)-1-Benzyl-3-aminopyrrolidine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only