Learn More
(S)-(-)-1-BOC-3-aminopyrrolidine, 95%, Thermo Scientific Chemicals
£87.80 - £309.00
Chemical Identifiers
CAS | 147081-44-5 |
---|---|
Molecular Formula | C9H18N2O2 |
Molecular Weight (g/mol) | 186.26 |
MDL Number | MFCD03419271 |
InChI Key | CMIBWIAICVBURI-UHFFFAOYNA-N |
Synonym | s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine |
PubChem CID | 854071 |
IUPAC Name | tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCC(N)C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10637030
|
Thermo Scientific Acros
435450010 |
1 g | Glass bottle |
£87.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783342
|
Thermo Scientific Acros
435450050 |
5 g | Glass bottle |
£309.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
147081-44-5 | |
186.26 | |
CMIBWIAICVBURI-UHFFFAOYNA-N | |
854071 | |
CC(C)(C)OC(=O)N1CCC(N)C1 |
C9H18N2O2 | |
MFCD03419271 | |
s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine | |
tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate |
Specifications
147081-44-5 | |
100.0 | |
120.0°C to 125.0°C (5.0 Torr) | |
Authentic | |
C9H18N2O2 | |
-1.5° to -3.0° (20°C, 589nm) (C=1, chloroform) | |
CMIBWIAICVBURI-UHFFFAOYNA-N | |
tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate | |
854071 | |
95% | |
(S)-(-)-1-BOC-3-aminopyrrolidine |
94.0 | |
Colorless to Yellow | |
91°C | |
94% min. (GC) | |
MFCD03419271 | |
s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine | |
CC(C)(C)OC(=O)N1CCC(N)C1 | |
186.26 | |
186.25 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Risk of explosion if heated under confinement.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
- (S)-(-)-1-Boc-3-aminopyrrolidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H301-Toxic if swallowed.
- EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only