Learn More
(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific Chemicals
£80.90 - £232.00
Chemical Identifiers
CAS | 195604-39-8 |
---|---|
Molecular Formula | C5H11N |
Molecular Weight (g/mol) | 85.15 |
MDL Number | MFCD08064289 |
InChI Key | IXCXVGWKYIDNOS-BYPYZUCNSA-N |
PubChem CID | 7363995 |
IUPAC Name | (1S)-1-cyclopropylethanamine |
SMILES | CC(C1CC1)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491740
|
Thermo Scientific Alfa Aesar
H27499.03 |
1 g |
£80.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401750
|
Thermo Scientific Alfa Aesar
H27499.06 |
5 g |
£232.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with acids and strong oxidizing agents.
Chemical Identifiers
195604-39-8 | |
85.15 | |
IXCXVGWKYIDNOS-BYPYZUCNSA-N | |
(1S)-1-cyclopropylethanamine |
C5H11N | |
MFCD08064289 | |
7363995 | |
CC(C1CC1)N |
Specifications
195604-39-8 | |
0.799 | |
3°C (37°F) | |
MFCD08064289 | |
Air Sensitive | |
Miscible with water. | |
CC(C1CC1)N | |
85.15 | |
85.15 | |
ee ≥98% |
-68°C | |
95°C | |
C5H11N | |
UN2733 | |
s-1-cyclopropylethylamine, s-1-cyclopropylethanamine, 1s-1-cyclopropylethanamine, 1s-1-cyclopropylethan-1-amine, 1 s-1-cyclopropylethanamine, n-s-1-cyclopropyl-ethyl-amine, s-1-cyclopropylethylamine, chipros, cyclopropanemethanamine,a-methyl-, as, s-1-cyclopropylethylamine, chipros, ee 98+%, s-1-cyclopropylethylamine, chipros r , produced by basf | |
IXCXVGWKYIDNOS-BYPYZUCNSA-N | |
(1S)-1-cyclopropylethanamine | |
7363995 | |
98% | |
(S)-1-Cyclopropylethylamine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only