missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Phenylbutylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific™

Product Code. 11406090
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3789-60-4
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD08064291
InChI Key XHOXKVFLASIOJD-JTQLQIEISA-N
Synonym s-1-phenylbutylamine, s-1-phenylbutan-1-amine, 1s-1-phenylbutan-1-amine, pubchem24143, s-alpha-phenylbutylamine, s-1-phenyl-butylamine, s-1-phenyl-1-butanamine, s-1-phenylbutylamine, chipros, benzenemethanamine, a-propyl-, as, s-1-phenylbutylamine, chipros , ee 98+%
PubChem CID 2449444
IUPAC Name (1S)-1-phenylbutan-1-amine
SMILES CCCC(C1=CC=CC=C1)N

Specifications

Boiling Point 220°C
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 149.24
Percent Purity 98%
Assay ee ≥98%
Chemical Name or Material (S)-1-Phenylbutylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'