missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals

Product Code. 15495495
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1365531-90-3
Molecular Formula C48H52O4P2
Molecular Weight (g/mol) 754.888
MDL Number MFCD19443624
InChI Key IDNKFBDRZBLTFQ-UHFFFAOYSA-N
Synonym 2'-bis 3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 3,5-dimethylphenyl phosphane, 2,2'-di di 3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-dimethylphenyl phosphine, r-2,2'-bis bis 3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxy--1,1'-biphenyl, r-2,2'-bis bis 3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-2,2'-bis bis 3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-dimethylphenyl phosphine
PubChem CID 87110781
IUPAC Name [2-[2-bis(3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1)P(C2=CC(=CC(=C2)C)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=CC(=C5)C)C)C6=CC(=CC(=C6)C)C)OC)OC)OC)OC)C

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 754.87
Percent Purity ≥97%
Chemical Name or Material (S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'