missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
0.5 g
2 g
10 g
Description
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl is chiral ligand widely used in asymmetric synthesis. It also used as excellent catalysts for asymmetric hydrogenation of alkenes and some cyclic anhydrides. BINAP is used in organic synthesis for enantioselective transformations catalyzed by its complexes of ruthenium, rhodium, and palladium
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 255°C to 257°C |
Quantity | 0.5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 678.79 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | (S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'