missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals
£93.63 - £339.00
Chemical Identifiers
CAS | 1365531-82-3 |
---|---|
Molecular Formula | C44H44O4P2 |
Molecular Weight (g/mol) | 698.78 |
MDL Number | MFCD19443623 |
InChI Key | JXFDRZQHVPTIMH-UHFFFAOYSA-N |
Synonym | s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine |
PubChem CID | 87110704 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436106
|
Thermo Scientific Alfa Aesar
H60921.MD |
250 mg | N/A | ||||||
15426106
|
Thermo Scientific Alfa Aesar
H60921.03 |
1 g |
£339.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1365531-82-3 | |
698.78 | |
JXFDRZQHVPTIMH-UHFFFAOYSA-N | |
87110704 |
C44H44O4P2 | |
MFCD19443623 | |
s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine |
Specifications
1365531-82-3 | |
MFCD19443623 | |
s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine | |
698.78 | |
698.77 | |
(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl |
C44H44O4P2 | |
Air sensitive | |
JXFDRZQHVPTIMH-UHFFFAOYSA-N | |
87110704 | |
≥97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only