missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol, 98%, Thermo Scientific Chemicals
£63.01 - £214.07
Chemical Identifiers
CAS | 22323-82-6 |
---|---|
Molecular Formula | C6H12O3 |
Molecular Weight (g/mol) | 132.16 |
MDL Number | MFCD00063239 |
InChI Key | RNVYQYLELCKWAN-UHFFFAOYNA-N |
Synonym | s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol |
PubChem CID | 736057 |
IUPAC Name | [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
SMILES | CC1(C)OCC(CO)O1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10421261
|
Thermo Scientific Acros
218070050 |
5 g | Glass bottle |
£63.01
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10033863
|
Thermo Scientific Acros
218070250 |
25 g | Glass bottle |
£214.07
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22323-82-6 | |
132.16 | |
RNVYQYLELCKWAN-UHFFFAOYNA-N | |
736057 | |
CC1(C)OCC(CO)O1 |
C6H12O3 | |
MFCD00063239 | |
s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol | |
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
Specifications
22323-82-6 | |
100.0 | |
1.0700g/mL | |
80°C | |
97.5% min. (GC) | |
1.4330 to 1.4350 | |
19,III,647 | |
1.07 | |
+ 11.00 | |
Solubility in water: miscible | |
CC1(C)OCC(CO)O1 | |
132.16 | |
132.16 | |
Liquid |
97.5 | |
Colorless to Yellow | |
82.0°C to 83.0°C (14.0 mmHg) | |
Authentic | |
C6H12O3 | |
MFCD00063239 | |
10,3236 | |
+ 11.00 (20.00°C c=5,CH3OH) | |
s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol | |
RNVYQYLELCKWAN-UHFFFAOYNA-N | |
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol | |
736057 | |
98% | |
(S)-(+)-2, 2-Dimethyl-1, 3-dioxolane-4-methanol |
Safety and Handling
missing translation for 'einecsNumber' : 244-910-8
Product Identifier
- (S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol
RUO – Research Use Only